Anti-SMO Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-2801R-A555
BOSSBS-2801R-A555EA 901 EUR
BOSSBS-2801R-A555
Anti-SMO Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
G protein-coupled receptor that probably associates with the patched protein (PTCH) to transduce the hedgehog's proteins signal. Binding of sonic hedgehog (SHH) to its receptor patched is thought to prevent normal inhibition by patched of smoothened (SMO). Required for the accumulation of KIF7 and GLI3 in the cilia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SMO
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR