Anti-Smoothened homolog Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-2801R-A680
BOSSBS-2801R-A680EA 899 EUR
BOSSBS-2801R-A680
Anti-Smoothened homolog Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
G protein-coupled receptor that probably associates with the patched protein (PTCH) to transduce the hedgehog's proteins signal. Binding of sonic hedgehog (SHH) to its receptor patched is thought to prevent normal inhibition by patched of smoothened (SMO). Required for the accumulation of KIF7 and GLI3 in the cilia.

Type: Primary
Antigen: Smoothened homolog
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 551-650/787
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR