Anti-SMO Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-2801R-CY5.5
BOSSBS-2801R-CY5.5EA 909 EUR
BOSSBS-2801R-CY5.5
Anti-SMO Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
G protein-coupled receptor that probably associates with the patched protein (PTCH) to transduce the hedgehog's proteins signal. Binding of sonic hedgehog (SHH) to its receptor patched is thought to prevent normal inhibition by patched of smoothened (SMO). Required for the accumulation of KIF7 and GLI3 in the cilia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SMO
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR