Anti-ARHGEF7 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-2936R-CY5
BOSSBS-2936R-CY5EA 900 EUR
BOSSBS-2936R-CY5
Anti-ARHGEF7 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
ARHGEF7 is a p21-activated protein kinase (Pak)-interacting exchange factor that has been identified as a putative guanine nucleotide exchange factor (GEF) for Rac/Cdc42. It is expressed ubiquitously in all the tissue examined and other isoforms are expressed mainly in the central nervous system. Regulated phosphorylation of ARHGEF7 is necessary to maintain the balance between normal signalling by EGFR and Src versus aberrant growth and transformation.Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene, but some of their full-length sequences have not been determined.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ARHGEF7
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat