Anti-FUCA1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-2940R-CY5
BOSSBS-2940R-CY5EA 901 EUR
BOSSBS-2940R-CY5
Anti-FUCA1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Alpha-L-fucosidase is responsible for hydrolyzing the alpha-1,6-linked fucose joined to the reducing-end N-acetylglucosamine of the carbohydrate moieties of glycoproteins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FUCA1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR