Anti-codA Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-2950R-A488
BOSSBS-2950R-A488EA 893 EUR
BOSSBS-2950R-A488
Anti-codA Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Converts cytosine to uracil or 5-methylcytosine to thymine by deaminating carbon number 4.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Cytosine deaminase
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Yeast
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR