Anti-codA Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-2950R-A555
BOSSBS-2950R-A555EA 893 EUR
BOSSBS-2950R-A555
Anti-codA Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Converts cytosine to uracil or 5-methylcytosine to thymine by deaminating carbon number 4.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Cytosine deaminase
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Yeast