Anti-Cytosine deaminase Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-2950R-A750
BOSSBS-2950R-A750EA 890 EUR
BOSSBS-2950R-A750
Anti-Cytosine deaminase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Converts cytosine to uracil or 5-methylcytosine to thymine by deaminating carbon number 4.

Type: Primary
Antigen: Cytosine deaminase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-158/158
Host: Rabbit
Isotype: IgG
Reactivity: Yeast
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR