Anti-codA Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-2950R-CY5.5
BOSSBS-2950R-CY5.5EA 900 EUR
BOSSBS-2950R-CY5.5
Anti-codA Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Converts cytosine to uracil or 5-methylcytosine to thymine by deaminating carbon number 4.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Cytosine deaminase
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Yeast