Anti-KCNK3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-2963R-A350
BOSSBS-2963R-A350EA 893 EUR
BOSSBS-2963R-A350
Anti-KCNK3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
This gene encodes a member of the superfamily of potassium channel proteins that contain two pore-forming P domains. The encoded protein is an outwardly rectifying channel that is sensitive to changes in extracellular pH and is inhibited by extracellular acidification. Also referred to as an acid-sensitive potassium channel, it is activated by the anesthetics halothane and isoflurane. Although three transcripts are detected in northern blots, there is currently no sequence available to confirm transcript variants for this gene.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KCNK3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat