Anti-KCNK3 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-2963R-CY7
BOSSBS-2963R-CY7EA 900 EUR
BOSSBS-2963R-CY7
Anti-KCNK3 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
This gene encodes a member of the superfamily of potassium channel proteins that contain two pore-forming P domains. The encoded protein is an outwardly rectifying channel that is sensitive to changes in extracellular pH and is inhibited by extracellular acidification. Also referred to as an acid-sensitive potassium channel, it is activated by the anesthetics halothane and isoflurane. Although three transcripts are detected in northern blots, there is currently no sequence available to confirm transcript variants for this gene.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KCNK3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat