Anti-MBP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-2967R-HRP
BOSSBS-2967R-HRPEA 371 EUR
BOSSBS-2967R-HRP
Anti-MBP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Involved in the high-affinity maltose membrane transport system MalEFGK. Initial receptor for the active transport of and chemotaxis toward maltooligosaccharides.

Recommended Dilutions: Western Blot: 1:100-1000

Type: Primary
Antigen: MBP
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR