Anti-TAGL2 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-2977R
BOSSBS-2977REA 725 EUR
BOSSBS-2977R
Anti-TAGL2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TAGL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat