Anti-TAGL2 Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-2977R-CY5.5
BOSSBS-2977R-CY5.5EA 900 EUR
BOSSBS-2977R-CY5.5
Anti-TAGL2 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Calponin homology domain; actin-binding domain which may be present as a single copy or in tandem repeats (which increases binding affinity). The CH domain is found in cytoskeletal and signal transduction proteins, including actin-binding proteins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TAGL2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat