Anti-FUS Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-2980R
BOSSBS-2980REA 725 EUR
BOSSBS-2980R
Anti-FUS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FUS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat