Anti-FUS Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-2980R-A488
BOSSBS-2980R-A488EA 893 EUR
BOSSBS-2980R-A488
Anti-FUS Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Binds both single-stranded and double-stranded DNA and promotes ATP-independent annealing of complementary single-stranded DNAs and D-loop formation in superhelical double-stranded DNA. May play a role in maintenance of genomic integrity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FUS
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR