Anti-FUS Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-2980R-A680
BOSSBS-2980R-A680EA 890 EUR
BOSSBS-2980R-A680
Anti-FUS Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Binds both single-stranded and double-stranded DNA and promotes ATP-independent annealing of complementary single-stranded DNAs and D-loop formation in superhelical double-stranded DNA. May play a role in maintenance of genomic integrity.

Type: Primary
Antigen: FUS
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 421-526/526
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR