Anti-FUS Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-2980R-A750
BOSSBS-2980R-A750EA 890 EUR
BOSSBS-2980R-A750
Anti-FUS Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Binds both single-stranded and double-stranded DNA and promotes ATP-independent annealing of complementary single-stranded DNAs and D-loop formation in superhelical double-stranded DNA. May play a role in maintenance of genomic integrity.
Type: Primary
Antigen: FUS
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 421-526/526
Host: Rabbit
Isotype: IgG
Reactivity: Rat