Anti-FUS Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-2980R-CY5
BOSSBS-2980R-CY5EA 900 EUR
BOSSBS-2980R-CY5
Anti-FUS Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Binds both single-stranded and double-stranded DNA and promotes ATP-independent annealing of complementary single-stranded DNAs and D-loop formation in superhelical double-stranded DNA. May play a role in maintenance of genomic integrity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FUS
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat