Anti-FUS Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-2980R-CY7
BOSSBS-2980R-CY7EA 900 EUR
BOSSBS-2980R-CY7
Anti-FUS Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Binds both single-stranded and double-stranded DNA and promotes ATP-independent annealing of complementary single-stranded DNAs and D-loop formation in superhelical double-stranded DNA. May play a role in maintenance of genomic integrity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FUS
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR