Anti-CYP2D6 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-2991R
BOSSBS-2991REA 704 EUR
BOSSBS-2991R
Anti-CYP2D6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYP2D6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen