Anti-CYP2D6 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-2991R-CY3
BOSSBS-2991R-CY3EA 883 EUR
BOSSBS-2991R-CY3
Anti-CYP2D6 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Responsible for the metabolism of many drugs and environmental chemicals that it oxidizes. It is involved in the metabolism of drugs such as antiarrhythmics, adrenoceptor antagonists, and tricyclic antidepressants.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP2D6
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR