Anti-BMX Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-3132R-A488
BOSSBS-3132R-A488EA 921 EUR
BOSSBS-3132R-A488
Anti-BMX Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
This gene encodes a non-receptor tyrosine kinase belonging to the Tec kinase family. The protein contains a PH-like domain, which mediates membrane targeting by binding to phosphatidylinositol 3,4,5-triphosphate (PIP3), and a SH2 domain that binds to tyrosine-phosphorylated proteins and functions in signal transduction. The protein is implicated in several signal transduction pathways including the Stat pathway, and regulates differentiation and tumorigenicity of several types of cancer cells. Multiple alternatively spliced variants, encoding the same protein, have been identified.[provided by RefSeq, Sep 2009].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BMX
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR