Anti-BMX Tyr40 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-3132R-A750
BOSSBS-3132R-A750EA 917 EUR
BOSSBS-3132R-A750
Anti-BMX Tyr40 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
This gene encodes a non-receptor tyrosine kinase belonging to the Tec kinase family. The protein contains a PH-like domain, which mediates membrane targeting by binding to phosphatidylinositol 3,4,5-triphosphate (PIP3), and a SH2 domain that binds to tyrosine-phosphorylated proteins and functions in signal transduction. The protein is implicated in several signal transduction pathways including the Stat pathway, and regulates differentiation and tumorigenicity of several types of cancer cells. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Type: Primary
Antigen: BMX Tyr40
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Rat