Anti-GAB2 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-3177R
BOSSBS-3177REA 743 EUR
BOSSBS-3177R
Anti-GAB2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GAB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat