Anti-NFKBIB Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-3191R-CY5
BOSSBS-3191R-CY5EA 929 EUR
BOSSBS-3191R-CY5
Anti-NFKBIB Rabbit Polyclonal Antibody (Cy5®)
Antikörper
The protein encoded by this gene belongs to the NF-kappa-B inhibitor family, which inhibit NF-kappa-B by complexing with, and trapping it in the cytoplasm. Phosphorylation of serine residues on these proteins by kinases marks them for destruction via the ubiquitination pathway, thereby allowing activation of the NF-kappa-B, which translocates to the nucleus to function as a transcription factor. Alternatively spliced transcript variants have been found for this gene.[provided by RefSeq, Jul 2011].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IKB beta
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat