Anti-STK11 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-3248R
BOSSBS-3248REA 743 EUR
BOSSBS-3248R
Anti-STK11 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STK11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR