Anti-LRP6 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-3253R
BOSSBS-3253REA 745 EUR
BOSSBS-3253R
Anti-LRP6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LRP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse