Anti-MARCKS Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-3263R
BOSSBS-3263REA 737 EUR
BOSSBS-3263R
Anti-MARCKS Rabbit Polyclonal Antibody
Antikörper
MARCKS, (Myristoylated Alanine-Rich C Kinase Substrate), is a member of a family of calmodulin binding proteins and is a major substrate for phosphorylation by protein kinase C (PKC). The phosphorylation of Ser152/156 can be used as a measure of PKC activation. Phosphorylation of Ser152/156 modulates the binding of MARCKS to calmodulin.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: MARCKS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat