Anti-MARCKS Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-3263R-CY5
BOSSBS-3263R-CY5EA 921 EUR
BOSSBS-3263R-CY5
Anti-MARCKS Rabbit Polyclonal Antibody (Cy5®)
Antikörper
MARCKS, (Myristoylated Alanine-Rich C Kinase Substrate), is a member of a family of calmodulin binding proteins and is a major substrate for phosphorylation by protein kinase C (PKC). The phosphorylation of Ser152/156 can be used as a measure of PKC activation. Phosphorylation of Ser152/156 modulates the binding of MARCKS to calmodulin.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MARCKS
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat