Anti-MST1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-3294R-A555
BOSSBS-3294R-A555EA 921 EUR
BOSSBS-3294R-A555
Anti-MST1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Protein which catalyzes the phosphorylation of serine or threonine residues on target proteins by using ATP as phosphate donor. Such phosphorylation may cause changes in the function of the target protein. Protein kinases share a conserved catalytic core common to both serine/ threonine and tyrosine protein kinases.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MST1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR