Anti-MST1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-3294R-CY5.5
BOSSBS-3294R-CY5.5EA 921 EUR
BOSSBS-3294R-CY5.5
Anti-MST1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Protein which catalyzes the phosphorylation of serine or threonine residues on target proteins by using ATP as phosphate donor. Such phosphorylation may cause changes in the function of the target protein. Protein kinases share a conserved catalytic core common to both serine/ threonine and tyrosine protein kinases.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MST1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

TECHNISCHE DATEN

Antigen-Symbol MST1
Antigen-Name Macrophage Stimulating 1
Environmentally Preferable
Antigen-Synonyme D3F15S2|STE20-like kinase MST1|Serine/threonine protein kinase 4|MSP|HGFL|Serine/threonine protein kinase Krs-2|DNF15S2|Mammalian STE20-like protein kinase 1|NF15S2
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Cy5.5®
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 6789, 6788
Isotyp IgG
ImmunoChemie Yes
Modification phospho-Thr 183/Thr180
Immunogen 150-200/487
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR