Anti-MST1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-3294R-CY7
BOSSBS-3294R-CY7EA 921 EUR
BOSSBS-3294R-CY7
Anti-MST1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Protein which catalyzes the phosphorylation of serine or threonine residues on target proteins by using ATP as phosphate donor. Such phosphorylation may cause changes in the function of the target protein. Protein kinases share a conserved catalytic core common to both serine/ threonine and tyrosine protein kinases.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MST1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR