Anti-GRIN1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-3301R-A555
BOSSBS-3301R-A555EA 921 EUR
BOSSBS-3301R-A555
Anti-GRIN1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
NMDA receptor subtype of glutamate-gated ion channels with high calcium permeability and voltage-dependent sensitivity to magnesium. Mediated by glycine. This protein plays a key role in synaptic plasticity, synaptogenesis, excitotoxicity, memory acquisition and learning. It mediates neuronal functions in glutamate neurotransmission. Is involved in the cell surface targeting of NMDA receptors (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NMDAR1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR