Anti-GRIN1 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-3301R-CY3
BOSSBS-3301R-CY3EA 929 EUR
BOSSBS-3301R-CY3
Anti-GRIN1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
NMDA receptor subtype of glutamate-gated ion channels with high calcium permeability and voltage-dependent sensitivity to magnesium. Mediated by glycine. This protein plays a key role in synaptic plasticity, synaptogenesis, excitotoxicity, memory acquisition and learning. It mediates neuronal functions in glutamate neurotransmission. Is involved in the cell surface targeting of NMDA receptors (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NMDAR1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat