Anti-GRIN1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-3302R-CY7
BOSSBS-3302R-CY7EA 929 EUR
BOSSBS-3302R-CY7
Anti-GRIN1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Neuronal Marker The protein encoded by this gene is a critical subunit of N-methyl-D-aspartate receptors, members of the glutamate receptor channel superfamily which are heteromeric protein complexes with multiple subunits arranged to form a ligand-gated ion channel. These subunits play a key role in the plasticity of synapses, which is believed to underlie memory and learning. Cell-specific factors are thought to control expression of different isoforms, possibly contributing to the functional diversity of the subunits. Alternatively spliced transcript variants have been described. [provided by RefSeq, Jul 2008]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NMDAR1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR