Anti-PLC gamma 2 Tyr759 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-3339R-A750
BOSSBS-3339R-A750EA 917 EUR
BOSSBS-3339R-A750
Anti-PLC gamma 2 Tyr759 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The protein encoded by this gene is a transmembrane signaling enzyme that catalyses the conversion of 1-phosphatidyl-1D-myo-inositol 4,5-bisphosphate to 1D-myo-inositol 1,4,5-trisphosphate (IP3) and diacylglycerol (DAG), using calcium as a cofactor. IP3 and DAG are second messenger molecules important for transmitting signals from growth factor receptors and immune system receptors across the cell membrane.
Type: Primary
Antigen: PLC gamma 2 Tyr759
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human