Anti-PLCG2 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-3339R-CY5
BOSSBS-3339R-CY5EA 929 EUR
BOSSBS-3339R-CY5
Anti-PLCG2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
The protein encoded by this gene is a transmembrane signaling enzyme that catalyzes the conversion of 1-phosphatidyl-1D-myo-inositol 4,5-bisphosphate to 1D-myo-inositol 1,4,5-trisphosphate (IP3) and diacylglycerol (DAG), using calcium as a cofactor. IP3 and DAG are second messenger molecules important for transmitting signals from growth factor receptors and immune system receptors across the cell membrane. [provided by RefSeq, Sep 2011].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PLCG2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat