Anti-PLCG2 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-3339R-CY5.5
BOSSBS-3339R-CY5.5EA 929 EUR
BOSSBS-3339R-CY5.5
Anti-PLCG2 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
The protein encoded by this gene is a transmembrane signaling enzyme that catalyzes the conversion of 1-phosphatidyl-1D-myo-inositol 4,5-bisphosphate to 1D-myo-inositol 1,4,5-trisphosphate (IP3) and diacylglycerol (DAG), using calcium as a cofactor. IP3 and DAG are second messenger molecules important for transmitting signals from growth factor receptors and immune system receptors across the cell membrane. [provided by RefSeq, Sep 2011].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PLCG2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR