Anti-DLG4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-3349R-A555
BOSSBS-3349R-A555EA 921 EUR
BOSSBS-3349R-A555
Anti-DLG4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
postsynaptic density protein 95 is a molecular scaffolding protein that binds and clusters N-methyl-D-aspartate receptors at neuronal synapses; may be involved in guanine nucleotide-mediated signal transduction pathway. This gene encodes a member of the membrane-associated guanylate kinase (MAGUK) family. It heteromultimerizes with DLG2. With DLG2 it is recruited into the same NMDA receptor and potassium channel clusters. These two MAGUK proteins may interact at postsynaptic sites to form a multimeric scaffold for the clustering of receptors, ion channels, and associated signaling proteins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DLG4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat