Anti-PSD95 Tyr236 + Tyr240 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-3349R-A750
BOSSBS-3349R-A750EA 917 EUR
BOSSBS-3349R-A750
Anti-PSD95 Tyr236 + Tyr240 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
postsynaptic density protein 95 is a molecular scaffolding protein that binds and clusters N-methyl-D-aspartate receptors at neuronal synapses; may be involved in guanine nucleotide-mediated signal transduction pathway. This gene encodes a member of the membrane-associated guanylate kinase (MAGUK) family. It heteromultimerizes with DLG2. With DLG2 it is recruited into the same NMDA receptor and potassium channel clusters. These two MAGUK proteins may interact at postsynaptic sites to form a multimeric scaffold for the clustering of receptors, ion channels, and associated signaling proteins.
Type: Primary
Antigen: PSD95 Tyr236 + Tyr240
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat