Anti-PIN1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-3354R-A647
BOSSBS-3354R-A647EA 1350 EUR
BOSSBS-3354R-A647
Anti-PIN1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Essential PPIase that regulates mitosis presumably by interacting with NIMA and attenuating its mitosis-promoting activity. Displays a preference for an acidic residue N-terminal to the isomerized proline bond. Catalyzes pSer/Thr-Pro cis/trans isomerizations. Down-regulates kinase activity of BTK. Can transactivate multiple oncogenes and induce centrosome amplification, chromosome instability and cell transformation. Required for the efficient dephosphorylation and recycling of RAF1 after mitogen activation. Binds and targets PML and BCL6 for degradation in a phosphorylation-dependent manner.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PIN1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR