Anti-MARCKS Ser162 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-3359R-A680
BOSSBS-3359R-A680EA 917 EUR
BOSSBS-3359R-A680
Anti-MARCKS Ser162 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
MARCKS, (Myristoylated Alanine-Rich C Kinase Substrate), is a member of a family of calmodulin binding proteins and is a major substrate for phosphorylation by protein kinase C (PKC). The phosphorylation of Ser152/156 can be used as a measure of PKC activation. Phosphorylation of Ser152/156 modulates the binding of MARCKS to calmodulin.
Type: Primary
Antigen: MARCKS Ser162
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen