Anti-TH Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-3463R-A555
BOSSBS-3463R-A555EA 921 EUR
BOSSBS-3463R-A555
Anti-TH Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Neuronal MarkerTyrosine hydroxylase is involved in the conversion of tyrosine to dopamine. As the rate-limiting enzyme in the synthesis of catecholamines, tyrosine hydroxylase has a key role in the physiology of adrenergic neurons. Tyrosine hydroxylase is regularly used as a marker for dopaminergic neurons, which is particularly relevant for research into Parkinson's disease.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR