Anti-TH Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-3463R-A647
BOSSBS-3463R-A647EA 921 EUR
BOSSBS-3463R-A647
Anti-TH Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Neuronal MarkerTyrosine hydroxylase is involved in the conversion of tyrosine to dopamine. As the rate-limiting enzyme in the synthesis of catecholamines, tyrosine hydroxylase has a key role in the physiology of adrenergic neurons. Tyrosine hydroxylase is regularly used as a marker for dopaminergic neurons, which is particularly relevant for research into Parkinson's disease.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat