Anti-NCF4 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-3497R-CY5.5
BOSSBS-3497R-CY5.5EA 929 EUR
BOSSBS-3497R-CY5.5
Anti-NCF4 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
NCF4 is a cytosolic oxidase associated protein. It is a component of the nicotinamide adenine dinucleotide phosphate oxidase, which mediates down regulation of NADPH oxidase through interactions with its SH3 domain. NCF4 is associated with p67 phox but is absent in patients with chronic granulomatous disease who lack p67 phox.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NCF4
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR