Anti-NCF4 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-3497R-CY7
BOSSBS-3497R-CY7EA 929 EUR
BOSSBS-3497R-CY7
Anti-NCF4 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
NCF4 is a cytosolic oxidase associated protein. It is a component of the nicotinamide adenine dinucleotide phosphate oxidase, which mediates down regulation of NADPH oxidase through interactions with its SH3 domain. NCF4 is associated with p67 phox but is absent in patients with chronic granulomatous disease who lack p67 phox.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NCF4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat