Anti-D-DIMER Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-3514R-A750
BOSSBS-3514R-A750EA 890 EUR
BOSSBS-3514R-A750
Anti-D-DIMER Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
D-DIMER is A fibrin degradation fragment or product that is produced by the action of plasmin on fibrin in the clot dissolution process.

Type: Primary
Antigen: D-DIMER
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR