Anti-PKC alpha/beta/gamma Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-3531R-A750
BOSSBS-3531R-A750EA 874 EUR
BOSSBS-3531R-A750
Anti-PKC alpha/beta/gamma Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Protein Kinase c alpha (PKC alpha) is an 77 kDa member of the conventional group (cPKCs: sensitive to calcium, diacylglycerol, phosphatidylserine and phorbol esters) of the PKC family of serine/ threonine kinases that are involved in a wide range of physiological processes including mitogenesis, cell survival and transcriptional regulation. PKC alpha is an ubiquitously expressed PKC isozyme that has been implicated in the regulation of a broad range of cellular functions including proliferation, differentiation, development, migration, cell cell adhesion, cell extracellular matrix adhesion, and solute transport. The activation loop threonine (threonine 497 in PKC alpha) of conventional PKCs is phosphorylated by phosphoinositide dependent kinase 1 (PDK1). This phosphorylation is necessary for the autophosphorylation of threonine 638 in the carboxy terminus of PKC alpha, a step that is critical for regulating the rate of PKC alpha dephosphorylation and inactivation.

Type: Primary
Antigen: PKC alpha/beta/gamma
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 151-250/672
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR