Anti-PPIG Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-3536R
BOSSBS-3536REA 725 EUR
BOSSBS-3536R
Anti-PPIG Rabbit Polyclonal Antibody
Antikörper
PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides. May be implicated in the folding, transport, and assembly of proteins. May play an important role in the regulation of pre-mRNA splicing.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: PPIG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR